Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Needle, Custom length, Point Style 2, 3, or 4
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
Brand: Hamilton™ 7748-09
This item is not returnable.
View return policy
Description
- Available in various gauges
- Autoclavable
- Metal hubs manufactured from nickel plated brass
- Custom length
- Compatible with PTFE Luer Lock (TLL) syringes
- Design controlled by a series of universal standards which guarantees compatibility between manufacturers
- Provided in packs of 6
Specifications
Autoclavable | |
23 G | |
0.64 mm | |
6/Pk. | |
TLL Syringes and LT or TLL Connectors |
Nickel-plated Brass (Hub), Stainless Steel | |
0.34 mm | |
Luer Lock Custom Needle | |
2, 3, or 4 | |
Standard Tapered |