Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Needle, Custom length, Point Style 2, 3, or 4
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
Brand:  Hamilton™ 7748-02
 View more versions of this product
Product Code. 11592335
  • 196.00 € / Pack of 6
Explore more special offers
Description
Description
  • Available in various gauges
  • Autoclavable
  • Metal hubs manufactured from nickel plated brass
  • Custom length
  • Compatible with PTFE Luer Lock (TLL) syringes
  • Design controlled by a series of universal standards which guarantees compatibility between manufacturers
  • Provided in packs of 6
Specifications
Specifications
Autoclavable
16 G
1.65 mm
6/Pk.
2, 3, or 4
Standard Tapered
Nickel-plated Brass (Hub), Stainless Steel
1.19 mm
Luer Lock Custom Needle
9.52 to 304 mm
TLL Syringes and LT or TLL Connectors