Learn More
Hamilton™ Custom Kel-F Hub Needles
Design controlled by a series of universal standards which guarantees compatibility between manufacturers.
Brand:  Hamilton™ 7750-16
 View more versions of this product
Product Code. 11502455
  • 200.00 € / Pack of 6
Explore more special offers
Description
Description
  • Available in various gauges
  • Provided in packs of 6
  • Custom point style
  • Autoclavable
  • Gas sterilizable
  • For Kel-F hubs
  • Custom length
  • Stainless steel Luer tip
Specifications
Specifications
Autoclavable
25 G
0.52 mm
6/Pk.
LT and TLL Syringes and Connectors
Stainless Steel
0.26 mm
Luer Lock Kel-F Hub Needle
2, 3, or 4
Standard Tapered