missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Custom Kel-F Hub Needles

Design controlled by a series of universal standards which guarantees compatibility between manufacturers.

Brand:  Hamilton™ 7750-21

 View more versions of this product

Product Code. 11532455

  • 200.00 € / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Available in various gauges
  • Provided in packs of 6
  • Custom point style
  • Autoclavable
  • Gas sterilizable
  • For Kel-F hubs
  • Custom length
  • Stainless steel Luer tip
Specifications

Specifications

Autoclavable
30 G
0.31 mm
6/Pk.
2, 3, or 4
Standard Tapered
Stainless Steel
0.16 mm
Luer Lock Kel-F Hub Needle
9.5 to 304 mm
LT and TLL Syringes and Connectors
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hamilton™ Custom Kel-F Hub Needles > Gauge: 30G; Needle Point Style: 2, 3, or 4

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.