missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Custom Kel-F Hub Needles
Design controlled by a series of universal standards which guarantees compatibility between manufacturers.
Brand: Hamilton™ 7750-05
This item is not returnable.
View return policy
Description
- Available in various gauges
- Provided in packs of 6
- Custom point style
- Autoclavable
- Gas sterilizable
- For Kel-F hubs
- Custom length
- Stainless steel Luer tip
Specifications
Autoclavable | |
14 G | |
2.11 mm | |
6/Pk. | |
LT and TLL Syringes and Connectors |
Stainless Steel | |
1.6 mm | |
Luer Lock Kel-F Hub Needle | |
Custom | |
Standard Tapered |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction