Learn More
Fisherbrand™ Stainless Steel Micro Chattaway Spatulas
Scrapes and lifts materials from tiny bottles or flasks. Fisherbrand™ Stainless Steel Chattaway Spatulas are non-magnetic with a bright finish.
Brand:  Fisherbrand™ 2350
 View more versions of this product
Product Code. 11523482
  • 32.40 € / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Manufactured from Stainless Steel
  • Bent and rounded end scrapes and lifts materials from bottles and flasks
  • Fits into tight flasks and bottles with necks ≥ 4mm diameter
Specifications
Specifications
5 Pk.
4 mm
100 x 4 mm
100 mm
Chattaway Spatula
1.6 mm (Barrel)
Stainless Steel