Learn More
Sampling Systems™ Stainless Steel Mini-Drums without Toggle Clamps
These Mini-Drums without Toggle Clamps are excellent multi-purpose containers for a wide variety of applications in the laboratory.
Brand:  Sampling Systems™ A434-1
 View more versions of this product
Product Code. 15899661
  • 263.00 € / Each
Explore more special offers
Description
Description
  • Manufactured from durable 304 grade stainless steel
  • Crevice-free interior
  • Neat recessed fitting lid
  • Complete with a swing carrying handle
  • Flat bottom with chime ring to give added protection
  • Available in a wide range of capacities to fit your specific needs
Specifications
Specifications
Yes
Recessed Lid
Multi-purpose Storage
Swing Handle
100 mm
Mini Drum
1 L
Stainless Steel
155 mm