Learn More
Sampling Systems™ Stainless Steel Mini Pots with Lids
Constructed of high quality stainless steel for durability. Sampling Systems™ Stainless Steel Mini Pots with Lids have a crevice free interior and are widely used in pharmaceutical and chemical laboratories.
41.80 € - 451.00 €
Specifications
Product Type Sample Jar
Material Stainless Steel (316L)
Closure Type Threaded Cap
Includes Lid and body
View More Specs
Products 8
Product Code Brand Capacity (Metric) Diameter (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Height (Metric) Price Quantity & Availability  
15819661
View Documents
Sampling Systems™
A426-100
100 mL 52 mm 73 mm
118.00 €
Each
15829661
View Documents
Sampling Systems™
A426-1000
1000 mL 102 mm 154 mm
451.00 €
Each
15839661
View Documents
Sampling Systems™
A426-25
25 mL 41 mm 36 mm
69.30 €
Each
15849661
View Documents
Sampling Systems™
A426-250
250 mL 77 mm 73 mm
207.00 €
Each
15859661
View Documents
Sampling Systems™
A426-50
50 mL 42 mm 53 mm
80.40 €
Each
15869661
View Documents
Sampling Systems™
A426-500
500 mL 102 mm 78 mm
301.00 €
Each
15879661
View Documents
Sampling Systems™
A426-7
7 mL 19 mm 45 mm
41.80 €
Each
15889661
View Documents
Sampling Systems™
A426-750
750 mL 102 mm 116 mm
318.00 €
Each
Description
Description
  • Available in a large range of sizes
  • Screw on lid
  • Satin finish
  • Multi purpose small volume containers
  • 316L Stainless Steel
  • Crevice free interior
  • Silicone O Ring available for improved sealing
Specifications
Specifications
Sample Jar
Stainless Steel (316L)
Threaded Cap
Lid and body