Learn More
Sampling Systems™ Stainless Steel Needle
This range of stainless steel needles makes light work of handling small and intricate materials.
Brand:  Sampling Systems™ A638-140
 View more versions of this product
Product Code. 17972012
  • 1.60 € / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Can be used to pick up small items or dissection
  • Available with either a straight end or curved end
  • Made from one piece and can be easily cleaned after use
  • Sharp point is ideal for use in laboratory and production work
Specifications
Specifications
Dissection Needle
Stainless Steel (AISI 304)
5.8 in.
Non-sterile
Silver
Straight
147 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sampling Systems™ Stainless Steel Needle > Stainless Steel Needle; Length (Metric): 147 mm; Style: Straight