Learn More
Sartorius™ Stainless Steel F1 Cylindrical Calibration Weight
Ensure accurate calibration and weighing. Fisherbrand™ Stainless Steel F1 Cylindrical Calibration Weight is a highly accurate individual weight with a nominal mass between 1g and 10kg. These cylindrical weights conform to class F1 requirements.
POA - 667.00 €
Specifications
Product Type | Cylindrical Calibration Weight |
---|---|
Classification | F1 |
Case Material | Plastic |
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
10563451
|
Sartorius™
YCW5138-00 |
100 g |
122.00 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
||||||
10441921
|
Sartorius™
YCW4138-00 |
10 g |
78.95 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||||
10034150
|
Sartorius™
YCW7138-00 |
10,000 g |
POA
Each |
N/A | ||||||
10553461
|
Sartorius™
YCW6138-00 |
1000 g |
293.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||||
10094140
|
Sartorius™
YCW3138-00 |
1 g |
68.30 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||||
10441531
|
Sartorius™
YCW5238-00 |
200 g |
150.00 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
||||||
10186510
|
Sartorius™
YCW3238-00 |
2 g |
69.30 €
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
10665951
|
Sartorius™
YCW4238-00 |
20 g |
85.30 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
||||||
10278410
|
Sartorius™
YCW6238-00 |
2000 g |
462.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||||
10656331
|
Sartorius™
YCW5538-00 |
500 g |
266.00 €
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
10655951
|
Sartorius™
YCW3538-00 |
5 g |
72.60 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
||||||
10145930
|
Sartorius™
YCW4538-00 |
50 g |
107.00 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
10145980
|
Sartorius™
YCW6538-00 |
5000 g |
667.00 €
Each |
Estimated Shipment: 11-04-2025
Log in to see stock. |
||||||
Description
The Fisherbrand™ line of reference line weights includes these cylindrical standard weights. They are constructed of durable stainless steel and conform to OIML class F1 for use with class II scales.- Available in a range of nominal mass values, from 2 to 10,000g
- Ideal for quality assurance in accordance with ISO 9001ff
- Adjustment, testing and calibration instruments for class II scales
- Quality assurance for scales with fewer than 300,000 steps
- Weights for calibration, and for official calibration of other weights, of the lower classes
(F2, F3, M1, M2 and M3)
Specifications
Cylindrical Calibration Weight | |
F1 | |
Plastic |