missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Essentials Stainless Steel Pail

This Pail is constructed of heavy gauge, 304 stainless steel and features a flat pouring rim.

Brand:  Cole-Parmer™ Essentials 07260-22

 View more versions of this product

Product Code. 11711836

  • 172.00 € / Each
Estimated Shipment: 27-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Minimize dripping with flat pouring rim
Specifications

Specifications

Pail
3.2 gal.
12 in.
304.8 mm
Stainless Steel
10 in.
12 in.
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Essentials Stainless Steel Pail > Stainless Steel Pail; Capacity: 12 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.