Learn More
Grant Instruments™ Stainless Steel PCR UV Cabinet
Disinfects the working area chamber, inactivating DNA / RNA fragments during 15-30 minutes of exposure. Grant Instruments™ Stainless Steel PCR UV Cabinet is equipped with a high efficiency flow-type bactericidal UV cleaner-recirculator which provide constant decontamination inside the cabinet during operation.
Brand:  Grant Instruments™ UVC/T-M-AR UK
2970.58 EUR valid until 2024-12-31
Use promo code "21853" to get your promotional price.
Product Code. 11441048
  • 3395.00 € / Each
Explore more special offers
Description
Description
  • Built-in patented UV cleaner-recirculator both increases the maximum density of UV light and generates 25 m³/h air flow exchange - prevents unwanted contamination and protects the user from direct UV light during manipulation
  • UV surface irradiation - via single 25W 254nm open UV lamp
  • High intensity UV cleaner - 25m³/hour cleaner-recirculator continuous air flow with 1cm UV irradiation distance
  • UV protection - UV-protective film on glass panels
  • UV exposure controlled by 24h digital timer
  • Convenient, easy to use digital timer for accurate control of UV exposure
  • White lamp provides local illumination of the workplace to optimise visual control during operations
  • Automatic swtich-off of irradiated UV light when door opened
  • Front opening with three adjustable positions for ease of access
Specifications
Specifications
15 W White Lamp
515 x 690 x 555 mm
120/230 V
Stainless Steel
50/60 Hz
31 kg