Learn More
Nickel Electro™ Stainless Steel Perforated Tray
Nickel Electro™ Stainless Steel Perforated Tray is to be used with flask clips and can be used in a variety of baths.
341.00 € - 544.00 €
Specifications
Material | Stainless Steel |
---|---|
For Use With (Equipment) | NE5-28 Shaker Baths |
Product Code | Brand | No. of Positions | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Positions | Product Type | Price | Quantity & Availability | ||||
12798968
|
Nickel Electro™
SFT0025 |
39 | Perforated Tray |
483.00 €
Each |
Estimated Shipment: 12-03-2025
Log in to see stock. |
||||
11736597
|
Nickel Electro™
SFT0050 |
32 | Perforated Tray |
472.00 €
Each |
Estimated Shipment: 12-03-2025
Log in to see stock. |
||||
12708978
|
Nickel Electro™
SFT0100 |
18 | Perforated Tray |
544.00 €
Each |
Estimated Shipment: 12-03-2025
Log in to see stock. |
||||
12718978
|
Nickel Electro™
SFT0250 |
12 | Perforated Tray |
341.00 €
Each |
Estimated Shipment: 12-03-2025
Log in to see stock. |
||||
12728978
|
Nickel Electro™
SFT0500 |
8 | Perforated Tray |
341.00 €
Each |
Estimated Shipment: 12-03-2025
Log in to see stock. |
||||
12738978
|
Nickel Electro™
SFT1000 |
5 | Perforated Tray |
341.00 €
Each |
Estimated Shipment: 12-03-2025
Log in to see stock. |
||||
12748978
|
Nickel Electro™
SFT2000 |
2 | Perforated Tray |
341.00 €
Each |
Estimated Shipment: 12-03-2025
Log in to see stock. |
||||
11419549
|
Nickel Electro™
SFT9000 |
2 x 1000 mL, 6 x 500 mL, 15 x 100 mL, 35 x 25 mL | Universal Spring Tray |
407.00 €
Each |
Estimated Shipment: 03-03-2025
Log in to see stock. |
||||
Description
- Supplied ready to assemble
- Stainless steel construction
- Suits glass/polypropylene Erlenmeyer flasks
- Tray simply drops onto shaker or rocking table platform and locates snugly
Specifications
Stainless Steel | |
NE5-28 Shaker Baths |