Learn More
Kap-Uts Culture Tube Closures
These Culture Tube Closures ensure the viability of microbiological cultures by protecting solid and broth media from excessive evaporation. They fit snugly on rimless culture tubes and flasks while allowing for gas exchange and ventilation.
720.00 € - 725.00 €
Specifications
Product Type Culture Tube Closure
Autoclavable Autoclavable
Material Stainless Steel
Quantity 144 Pk.
Tube Material Stainless Steel
View More Specs
Products 2
Product Code Brand Size (Metric) Price Quantity & Availability  
Product Code Brand Size (Metric) Price Quantity & Availability  
15245376
View Documents
Kap-Uts
04500-00
13 mm
725.00 €
Pack of 144
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
15215637
View Documents
Kap-Uts
04500-01
16 mm
720.00 €
Pack of 144
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
Description
Description
  • Closures minimize evaporation while allowing necessary gas exchange.
  • Positive-grip closures of stainless steel or autoclavable polypropylene.
Specifications
Specifications
Culture Tube Closure
Stainless Steel
Stainless Steel
Autoclavable
144 Pk.