Learn More
Grant Instruments™ Stainless Steel Perforated Base Tray
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
Brand:  Grant Instruments™ SBT12
 View more versions of this product
Product Code. 15315832
  • 125.08 € / Each
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
  • For use with SBB Aqua Plus and LSB Series Aqua Pro Series Unstirred Water Bath
  • Choose from a vartiety of sizes.
Specifications
Specifications
LSB12
SBT 12 Stainless Steel Perforated Base Tray
Stainless Steel