missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Stainless Steel Safety Cans

For safe metering and handling of smaller amounts of flammable liquids.Protection from exploding through flame protection and excess pressure valve, containers have a high degree of chemical resistance.

Brand:  Buerkle™ 2601-1002

465.14 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 11931551

  • 615.00 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless steel AISI 316 Ti (1.4571), 0.6 mm thickness
  • Seals made of PTFE
  • Handle made of powder-surfaced Aluminum
  • Closure thread 1¼in.
Specifications

Specifications

Safety Cans
For Transport and Storage
295 mm
2 L
Stainless Steel
131 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.