Learn More
Sampling Systems™ Stainless Steel Sample Splitter Riffle Boxes
These easy-to-use Sample Splitters in stainless steel are for dividing and reducing bulk samples.
Brand:  Sampling Systems™ 8601S-30
 View more versions of this product
Product Code. 15809541
  • 1385.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Simply drop the sample into the top and the sample is split in two
  • Split sample drops into the riffle boxes underneath
  • It is recommended that width of slots are 2.5 to 3 times bigger than the diameter of the largest particle being split
  • Available in a wide range of sizes
Specifications
Specifications
Riffle Box
12
30 mm
Stainless Steel
Dividing and reducing bulk samples