Learn More
Bel-Art™ SP Scienceware™ Stainless-steel Sampling Spoon and Spatula
Ends coated with brown Fluo-Kem™ PTFE FEP.
Brand:  Bel-Art™ H36711-0000
Product Code. 15530122
  • 47.55 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ends coated with brown Fluo-Kem™ PTFE FEP.
  • Coating provides anti-stick, corrosion-resistant surface
  • Rod flattened at one end to serve as spatula with spoon on opposite end
Specifications
Specifications
Spoon and Spatula Sampler
4mm dia. rod flattened at one end to serve as 8 x 51mm spatula with 13 x 25mm spoon on opposite end
22.9 cm
Stainless Steel
Brown Fluo-Kem PTFE FEP