Learn More
Swann-Morton™ Metal Diecast Handle, Postmortem Scalpel
Use the Swann Morton™ Metal Diecast Handle, Postmortem Scalpel for postmortem surgical techniques.
Brand:  Swann-Morton™ 2652
Product Code. 12922555
  • 112.00 € / Pack of 10
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Heavy duty carbon steel blade
  • Metal handle with screw-type fitting
Specifications
Specifications
Stainless Steel
Handle
PM40 or PM40 Bull Nose Pathology Blade
Epoxy Powder Coated
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products