Learn More
Kinematica™ Stainless Steel Sieve for Polymix™ MFC 90 D Mill
Depending on the sample, even finer results can be achieved by grinding with two or more sieves with progressively smaller hole sizes.
Brand:  Kinematica™ 35095030
 View more versions of this product
Product Code. 10387183
  • 170.00 € / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Sieve
PX MFC 90 D
Stainless Steel
3 mm