Learn More
Kinematica™ Stainless Steel Sieve for Polymix™ MFC 90 D Mill
Depending on the sample, even finer results can be achieved by grinding with two or more sieves with progressively smaller hole sizes.
Brand:  Kinematica™ 35095010
 View more versions of this product
Product Code. 10532274
  • 200.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Sieve
PX MFC 90 D
Stainless Steel
1 mm