Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SF55, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ SF55E02072
Product Code. 17538315
  • 87.15 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SF55 Sterilizer
Drip Tray
Rim: 15 mm
Stainless Steel