missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Stainless Steel Small Parts Cleaner

The Stainless Steel Small Parts Cleaner is ideal for cleaning or degreasing small parts.

Brand:  asecos™ 40446

 View more versions of this product

Product Code. 17826466

  • 158.00 € / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With recessed grip
  • Spring-loaded immersion sieve incl. flame arrester, grounding connection
Specifications

Specifications

Parts Cleaner
4 L
Steel
264.00 mm
146.00 mm
2.8 kg
1.1 gal.
Yellow
10.39 in.
5.75 in.
6.17 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.