missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stainless Steel Spatulas

Designed using high quality stainless steel and nickel components, these spatulas have good mechanical properties and excellent resistance to many corrosive environments.

Brand:  Fisherbrand™ 2262

Product Code. 11593452

  • 55.45 € / Pack of 5
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Spatulas, nickel and stainless steel Chattaway 3.6mm barrel, nickel and stainless micros 1.6mm barrel
  • Includes spoon on one end and curved blade on the other
Specifications

Specifications

5 Pk.
Spatula
Stainless Steel
165 mL
With a Spoon at One End and Curved Blade to Other
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.