missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Stainless-steel Spoons
Double-ended with two bowl sizes.
Brand: Bel-Art™ F36730-0015
This item is not returnable.
View return policy
Description
Double-ended with two bowl sizes.
- Made from 304 stainless-steel
- Sampling spoons have solid, rounded shaft
- Available in plain polished stainless-steel or coated with Fluo-Kem™ PTFE FEP
Specifications
Spoon | |
0.5, 2 mL | |
Brown | |
15 cm |
Stainless Steel | |
Solid, rounded shaft | |
Fluo-Kem Teflon FEP coated |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction