Learn More
Bel-Art™ SP Scienceware™ Stainless-steel Spoons
Double-ended with two bowl sizes.
Brand:  Bel-Art™ F36730-0030
 View more versions of this product
Product Code. 11726545
  • 157.99 € / Pack of 2
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Double-ended with two bowl sizes.
  • Made from 304 stainless-steel
  • Sampling spoons have solid, rounded shaft
  • Available in plain polished stainless-steel or coated with Fluo-Kem™ PTFE FEP
Specifications
Specifications
Spoon
Solid, rounded shaft
30 cm
2.0 mL small spoon, 8.0 mL large spoon
Fluo-Kem Teflon FEP coated ends