Learn More
Bel-Art™ SP Scienceware™ Stainless-steel Spoons
Double-ended with two bowl sizes.
Brand:  Bel-Art™ H36729-0025
 View more versions of this product
Product Code. 15570122
  • 108.00 € / Pack of 2
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Double-ended with two bowl sizes.
  • Made from 304 stainless-steel
  • Sampling spoons have solid, rounded shaft
  • Available in plain polished stainless-steel or coated with Fluo-Kem™ PTFE FEP
Specifications
Specifications
Spoon
Solid, rounded shaft
25 cm
1.8 mL small spoon, 5.0 mL large spoon
Plain-polished 304 grade stainless-steel