Learn More
asecos™ Stainless Steel Storage Can with Dosing Tap
The Stainless Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
163.00 € - 208.00 €
Specifications
Color Gray
Material Stainless Steel
View More Specs
Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
17876516
View Documents
asecos™
40084
1 L
163.00 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
17866476
View Documents
asecos™
40085
2 L
168.00 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
17886586
View Documents
asecos™
40086
5 L
208.00 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
Description
Description
  • With fine dosing tap made of stainless steel (1.4301), incl. vent, PTFE seal, integrated flame barrier (removable) and plastic protective ring
  • Incl. multilingual safety sticker to designate the contents
Specifications
Specifications
Gray
Stainless Steel