missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Stainless Steel Storage Can with Screw Cap
The Stainless Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
136.00 € - 179.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17856506
|
asecos™
40081 |
1 L |
136.00 €
Each |
Estimated Shipment: 03-03-2025
Log in to see stock. |
|||||
17856346
|
asecos™
40082 |
2 L |
146.00 €
Each |
Estimated Shipment: 03-03-2025
Log in to see stock. |
|||||
17826516
|
asecos™
40083 |
5 L |
179.00 €
Each |
Estimated Shipment: 03-03-2025
Log in to see stock. |
|||||
Description
- With screw cap (secured against loss) overpressure valve, PTFE seal, integrated flame barrier (removable) and plastic protective ring
- Incl. multilingual safety sticker to designate the contents
Specifications
Gray | |
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title