Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, in Micrometer Pore with ASTM E 11
Brand:  RETSCH 60.132.000600
 View more versions of this product
Product Code. 10084921
  • 121.00 € / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1 Each
Test Sieve
Shaker AS 200
50mm
200 mm
Stainless Steel
600 μm
ASTM E 11
Certificate indicating Compliance
For separation, fractioning, particle size determination
Stainless Steel