missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Stainless Steel Transport Canister

The Stainless Steel Transport Canister is ideal to safely transport of liquid substances of packing groups I, II and III.

156.00 € - 219.00 €

Specifications

Color Gray
Material Stainless Steel
View More Specs

Products 5
Product Code Brand Weight (Metric) Price Quantity & Availability  
Product Code Brand Weight (Metric) Price Quantity & Availability  
17806556
View Documents
asecos™
40087
2 kg
156.00 €
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
17816556
View Documents
asecos™
40088
2.2 kg
168.00 €
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
17836476
View Documents
asecos™
36480
2.75 kg
174.00 €
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
17856386
View Documents
asecos™
40089
3.3 kg
186.00 €
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
17886366
View Documents
asecos™
36481
3.8 kg
219.00 €
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
Description

Description

  • incl. multilingual safety sticker for content labelling, with aluminium screw cap, ventilation valve, NBR seals and flexible plastic outlet pipe (Ø approx. 20 mm)
  • UN-approval for packaging groups I, II and III
  • Stainless steel
Specifications

Specifications

Gray
Stainless Steel
Videos
SDS
Documents

Documents

Product Certifications