Learn More
JP Selecta™ Stainless Steel Tripod Support Stand
JP Selecta™ Tripod Support Rods is suitable for laboratory purposes to support tripod.
Brand:  JP Selecta™ 6002278
 View more versions of this product
Product Code. 12022305
  • 81.00 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of AISI 304 stainless steel
Specifications
Specifications
Stainless Steel
8 mm
1 Ea.
500 mm
Tripod support rod
1.4 kg