Learn More
JP Selecta™ Stainless Steel Tripod Support Stand
JP Selecta™ Tripod Support Rods is suitable for laboratory purposes to support tripod.
Brand:  JP Selecta™ 6022712
 View more versions of this product
Product Code. 12022365
  • 139.00 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of AISI 304 stainless steel
Specifications
Specifications
Stainless Steel
12 mm
1 Ea.
700 mm
Tripod support rod
2.5 kg