Learn More
Gilac™ Stainless Steel Trolley for Large Volume Container
Stainless Steel Trolley for Large Volume Container makes it easier to work at breast height. Equipped with 125 mm diameter wheels in white PA (2 fixed and 2 pivoting with brakes).
Brand:  Gilac™ G194895
 View more versions of this product
Product Code. 17958941
  • 1650.00 € / Each
Explore more special offers
Description
Description
  • Trolley equiped with 125 mm diameter wheels in white PA (2 fixed and 2 pivoting with brakes).
  • Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
  • Makes it easier to work at breast height.
Specifications
Specifications
Trolley
150 kg
125 mm
35.4 in.
Large Volume Containers
Stainless Steel
220 L Large Volume Semicircular Container
900 mm