Learn More
Lauda™ Stainless Steel Tubing Connectors
Lauda™ Stainless Steel Tubing Connectors are used for connecting polymer tubing to LAUDA thermostats and circulation chillers.
Brand:  Lauda™ HKO 025
Product Code. 10724703
  • 19.65 € / Each
Explore more special offers
Description
Description
  • For connecting polymer tubings
Specifications
Specifications
Tubing Connector
Stainless Steel
11 mm