Learn More
Lauda™ Stainless Steel Tubing Connectors
Lauda™ Stainless Steel Tubing Connectors are used for connecting polymer tubing to LAUDA thermostats and circulation chillers.
Brand:  Lauda™ HKO 061
Product Code. 10734703
  • 19.95 € / Each
Explore more special offers
Description
Description
  • For connecting polymer tubings
Specifications
Specifications
Tubing Connector
Stainless Steel
8.5 mm