Learn More
Sampling Systems™ Stainless Steel Tweezers
Stainless steel tweezers ideal for a variety of applications across a wide range of industries, including pharmaceutical. They are ideal for handling small items such as tissues and vials, and are made from medical grade stainless steel.
Brand:  Sampling Systems™ A394-400
Product Code. 17932102
  • 42.80 € / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made from 304 stainless steel
  • Available in a range of different lengths
  • Robust design
  • Useful for handling intricate items such as tissues
Specifications
Specifications
Serrated
Stainless Steel (304)
Tweezers
Non-sterile
15.7 in.
400 mm
1 Tweezer/Cs.
Standard
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sampling Systems™ Stainless Steel Tweezers > Stainless Steel Tweezers; Material: Stainless Steel (304)