Learn More
Cole-Parmer™ Stainless Steel Tweezers
Recommended for micro electronics and precision assembly
Brand:  Cole-Parmer™ 07287-08
 View more versions of this product
Product Code. 11705499
  • 18.20 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Recommended for micro electronics and precision assembly
  • Hand finished points for accuracy and excellent tip hardness
  • Anti-glare finish reduces eye strain
  • Anti-magnetic and acid resistant
Specifications
Specifications
Angled
4.25 in.
10.8 cm
Tweezer
Straight
Each
Stainless Steel
Tweezers
Ea.