Learn More
Thermo Scientific™ Stainless Steel Unions, Tees and Crosses
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Stainless-Steel Unions, Tees and Crosses.
Brand:  Thermo Scientific™ U-435
 View more versions of this product
Product Code. 10097214
  • 63.03 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ Stainless-Steel Unions, Tees and Crosses. Includes two stainless-steel nuts and ferrules.
  • Absolute zero or low dead volume formats
  • Includes two stainless steel nuts and ferrules
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Union
Union with 2 stainless steel nuts and ferrules
Stainless Steel
0.025 μL
0.010 in. through hole
Upchurch Scientific/Parker fittings
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.