missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Essentials Utility Bowls, 304 Stainless Steel
Heavy gauge, 304 stainless steel (SS) Utility Bowls with flat bottom and straight sides to ensure stability.
68.75 € - 89.55 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11797255
|
Cole-Parmer™ Essentials
07272-00 |
858 mL |
89.55 €
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
||||||
11707265
|
Cole-Parmer™ Essentials
07274-00 |
2000 mL |
68.75 €
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
Description
- These straight-sided, heavy-duty bowls are made of 304 SS with a satin finish that makes them easy to clean.
Specifications
Stainless Steel | |
Utility Bowl |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Essentials Utility Bowls, 304 Stainless Steel