missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Stainless Steel Utility Dipper

Sampling Systems™ Stainless Steel Utility Dipper is an all stainless steel sampler suitable for sampling liquids and suspensions.

169.00 € - 187.00 €

Specifications

Material Stainless Steel (AISI 304)
For Use With (Application) Low Viscosity Liquids
Diameter (Metric) 32 mm
View More Specs

Products 2
Product Code Brand Capacity (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Length (Metric) Price Quantity & Availability  
15809371
View Documents
Sampling Systems™
5170A-50
50 mL 460 mm
169.00 €
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
 
15899361
View Documents
Sampling Systems™
5170A-100
100 mL 540 mm
187.00 €
Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
Description

Description

  • Single point sampler
  • 304 Stainless Steel
  • Insert the sampler into the product for the liquid will flow into the sampler
  • Raise the centre control rod to discharge the sample
Specifications

Specifications

Stainless Steel (AISI 304)
Low Viscosity Liquids
32 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.