Learn More
Stainless-Steel wire, annealed, Thermo Scientific Chemicals
Temper: soft
22.10 € - 91.86 €
Specifications
Chemical Name or Material | Stainless Steel |
---|---|
Color | Silver |
Name Note | annealed, Type 304 |
Appearance | Silver-colored wire |
Molecular Formula | Fe:Cr:Ni; 70:19:11 wt% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11309448
|
Thermo Scientific Alfa Aesar
043301.BU |
25 cm | N/A | ||||||
11319448
|
Thermo Scientific Alfa Aesar
043301.BY |
100 cm |
45.63 €
100cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399438
|
Thermo Scientific Alfa Aesar
043301.CC |
500 cm |
91.86 €
500cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Stainless Steel Wire are suitable for making Kitchenware, Basket, Cable Trays and Trolleys with electro-polishing as the final process . Clean, de-greased & defect free surface results in very lustrous appearance when electro polished.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel | |
annealed, Type 304 | |
Fe:Cr:Ni; 70:19:11 wt% | |
Insoluble in water. | |
Wire |
Silver | |
Silver-colored wire | |
MFCD00148499 | |
Odorless |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.