Learn More
Tuttnauer Stainless Steel Wire Basket for Bench Top Autoclaves
Stainless steel mesh wire basket for bottles and general use.
Brand:  Tuttnauer BSK411-0005
 View more versions of this product
Product Code. 17879558
  • 295.00 € / Each
Explore more special offers
Description
Description
  • Stainless Steel wire basket
Specifications
Specifications
Maximum number of baskets: 1
11.5 L
7.4 in.
15.7 in.
Autoclave Basket
400 x 180 x 160 mm
2.5 gal.
18 cm
2840 Bench Top Autoclave
40 cm
Non-sterile