Learn More
Tuttnauer Stainless Steel Wire Basket for Vertical Autoclaves
Stainless steel mesh wire basket for bottles and general use.
Brand:  Tuttnauer BSK387-0001
 View more versions of this product
Product Code. 17889548
  • 362.00 € / Each
Explore more special offers
Description
Description
  • Stainless Steel wire basket
Specifications
Specifications
Maximum number of baskets: 2
39 L
357 x 330 mm
3850 Vertical Autoclave
35.7 cm
Non-sterile
8.6 gal.
33 cm
13.2 in.
14.1 in.
Autoclave Basket