missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Stainless Steel Wire Brushes

These durable stainless steel wire brushes with nylon bristles are available in a variety of sizes for cleaning parts, tubing and hard to reach areas.

Brand:  Micronova™ BRWM-125

 View more versions of this product

Product Code. 15783004

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Micronova Stainless Steel Wire Brushes are excellent for cleaning valves, tubing, cylinders, narrow chambers and other hard to clean areas. The durable brushes are flexible enough bend through fittings but stiff enough for removing residues. The tapered versions add a level of cleaning versatility.

  • Nylon bristle brushes with a stainless steel wire core
  • Ideal for valves, tubing, cylinders and narrow chambers
  • Flexible to bend through fittings but stiff enough to remove residues
  • Tapered versions add cleaning versatility
  • Available in a range of diameters from 0.25 to 2 inches
Specifications

Specifications

Brush
Stainless Steel Wire
Not Autoclavable
White
1.3 cm
Brush to Clean Tubing and Small Parts
40.6 cm
1.3 cm
Nylon
Non-sterile
Keep at ambient temperature
0.5 in.
Reusable
16 in.
0.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers