missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Worm-Threaded Hose Clips
Manufactured from stainless steel 1.4016.
Brand: Buerkle™ 8677-1220
17.67 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- With screw thread
- Screw is zinc-coated
Specifications
12 to 20 mm | |
Hoses | |
12 to 20 mm dia. |
Stainless Steel | |
10/Pk. | |
Worm-threaded Hose Clip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction