missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Worm-Threaded Hose Clips

Manufactured from stainless steel 1.4016.

Brand:  Buerkle™ 8677-1016

14.41 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10428151

  • 17.70 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With screw thread
  • Screw is zinc-coated
Specifications

Specifications

10 to 16 mm
Hoses
10 to 16 mm dia.
Stainless Steel
10/Pk.
Worm-threaded Hose Clip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.