Learn More
Bochem™ Stand Base with Adjustable Foot
Bochem™ steel varnished stand base has an adjustable foot for stability and is used with M10 rods, rings and clamps for supporting various laboratory apparatus.
Brand:  Bochem™ 5036
Product Code. 10578272
  • 44.65 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Steel varnished base DIN 12892
  • Thread M10
  • Adjustable foot
Specifications
Specifications
Varnished Steel
250 mm
Stand Base
Rods (Thread M10) and Clamps
250 x 160 mm
160 mm
Holding and Supporting Laboratory Apparatus