missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Standard Buffer Solutions pH 1.68, WTW™
With a high accuracy: ±0.02 pH, these standard buffering solutions by WTW are traceably certified to PTB/NIST and therefore safe for use.
Brand: WTW 109000
29.02 EUR valid until 2024-12-31
Use promo code "21980" to get your promotional price.
This item is not returnable.
View return policy
Description
- For precision applications
- PTB/NIST traceable standard buffer solutions (work reference buffer solutions) for precision applications - safe use
- Standard buffering solutions as per DIN 19266-01: 2000
- Universal buffering solutions
- High accuracy: ±0.02 pH
- Standard buffering solutions by WTW are traceably certified to PTB/NIST and therefore safe for use
- Buffer pH 1.679 and PL 2 are available in 250 ml bottles
Specifications
Colorless | |
Precision applications | |
Liquid | |
Bottle |
DIN/NIST | |
1.68 | |
250 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction