missing translation for 'onlineSavingsMsg'
Learn More

Standard Buffer Solution pH 4.01 and 6.87, SI Analytics™

Product Code. 10470071
Click to view available options
Quantity:
2 x 30 Ampules
Packaging:
FIOLAX™ Ampules
This item is not returnable. View return policy
This item is not returnable. View return policy

SI Analytics™ Standard Buffer Solution pH 4.01 and 6.87 resists changes in pH when small quantities of an acid or an alkali are added to it.

  • Includes manufacturer certificate
  • Mixture, consisting of the following components: Water, potassium hydrogenphthalate
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
Color Colorless
Density 1.01g/cm3
pH 4.01, 6.87
Physical Form Liquid
Quantity 2 x 30 Ampules
Formulation Water, potassium hydrogenphthalate
Odor Odorless
Packaging FIOLAX™ Ampules
Sterility Sterile
Vapor Pressure 23hPa
Solubility Information Fully miscible in water
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'